• USA Home
  • 37430 - AQUANAL-plus sulfide (S2-) 0.03-0.8 mg/L
EMAIL THIS PAGE TO A FRIEND
37430 Sigma
AQUANAL-plus sulfide (S2-) 0.03-0.8 mg/L
test set with color comparator
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items, Tests for semi-quantitative determination (color charts) More...
usage   sufficient for 100 tests
measuring range   0.03-0.8 mg/L, (color chart)
  0.05-1.0 mg/L, (photometric, Spectro 3)
Description
Analysis Note
Method
Formation of a blue dye with N,N-dimethyl-1,4-phenylenediamine dihydrochloride
Gradation (color chart)
0 - 0.03 - 0.06 - 0.10 - 0.15 - 0.20 - 0.30 - 0.40 - 0.60 - 0.80 mg/L (S2-)
Other Notes
Same product — same quality — now Sigma brand
General description
Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3316 9
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
replacement part
Product #
Description
Add to Cart
32022 AQUANAL-plus sulfide (S2-) 0.03-0.8 mg/L, refill pack for 37430
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?