• USA Home
  • 37500 - AQUANAL-plus cyanide/thiocyanate (CN/SCN) 0.03-0.7 mg/L

EMAIL THIS PAGE TO A FRIEND
37500 Sigma

AQUANAL-plus cyanide/thiocyanate (CN/SCN) 0.03-0.7 mg/L

check solution for 37420 (calibrated thiocyanate standard (CN/SCN = 0.005 mg/mL))

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
grade   check solution for 37420 (calibrated thiocyanate standard (CN/SCN = 0.005 mg/mL))

Description

General description

Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?