• USA Home
  • 38378 - Cellulose acetate sheets
EMAIL THIS PAGE TO A FRIEND
38378 Sigma
Cellulose acetate sheets
size 25 mm × 130 mm
Purchase
Properties
Related Categories Cellulose Electrophoresis, Molecular Biology, Protein Electrophoresis, Proteomics
size   25 mm × 130 mm
Description
Features and Benefits
With cellulose acetate electrophoresis, you can separate nearly all sample materials that migrate in an electrical field. Apply 0.06 to 0.15 mg of sample mixture per cm application line. Concentrate the mixture to at least 1 mg/ml. Advantages of those strips include:
• High porosity of the strip (approx. 80 % free pore volume)
• Isotropic, integral matrix for ideal flow conditions
• Minimal diffusion effect and low adsorption capacity
• Perfect transparency because of the pore system
• Ideal for the non-toxic ATX reagent system
• Virtually unlimited stability at room temperature
• Two identical sides, no need to search for the "right" side
Available in different sizes for all current chamber types
These strips can also be used for high-voltage electrophoresis, chromatography, and immunodiffusion.
Price and Availability
Detect Your Unseen Protein
Safety & Documentation
Safety Information
WGK Germany 
nwg
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?