EMAIL THIS PAGE TO A FRIEND
3861 Sigma

Citrate Concentrate

Purchase

Properties

Related Categories Cell Biology, Cytochemistry Products, Hematology and Histology More...
IVD   for in vitro diagnostic use
Quality Level   GMP-IVD
shipped in   wet ice
storage temp.   2-8°C

Description

Components

Citrate buffer, 0.38 mol/L, pH 5.4 when diluted according to procedure 386A

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?