EMAIL THIS PAGE TO A FRIEND
39335 Sigma

Biotin-amidite

≥96.0% (NMR)

Purchase

Properties

InChI Key   WSDQYLGSIUJJRU-ZMKGMYJVSA-N
assay   ≥96.0% (NMR)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?