Synonym: Detergent Screening Solution 35/Fluka kit no 66317, MEGA-8 solution
CAS Number 85316-98-9
MDL number MFCD00134152
PubChem Substance ID 24864627
| Related Categories | Detergents, Detergents A to M, Detergents by Applications, Diagnostic Applications, Drug Delivery-Liposomes, |
| description | non-ionic |
| CMC | 58 |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?