EMAIL THIS PAGE TO A FRIEND
396052 Sigma

o-Xylene

Purchase

Properties

storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1307 - class 3 - PG 3 - Xylenes
WGK Germany 
2
RTECS 
ZE2450000
Flash Point(F) 
87.8 °F
Flash Point(C) 
31 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?