• USA Home
  • 40601 - NeutrAvidin®-R-Phycoerythrin conjugate

EMAIL THIS PAGE TO A FRIEND
40601 Sigma

NeutrAvidin®-R-Phycoerythrin conjugate

neutrally charged avidin

Purchase

Properties

fluorescence   λex 565 nm; λem 576 nm in 0.1 M phosphate pH 7.0
storage temp.   2-8°C

Description

Application

NeutrAvidin-R-Phycoerythrin conjugate is a biotin-binding protein conjugated to the fluorescence indicator R-Phycoerythrin. NeutrAvidin-R-Phycoerythrin conjugate may be used in place of avidin or streptavidin R-Phycoerythrin conjugates to label biotin conjugated molecules in applications such as immunohistochemistry, DNA hybridization, MHC tetramer technology and flow cytometry.

Legal Information

NeutrAvidin is a registered trademark of Thermo Fisher Scientific, Inc.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?