• USA Home
  • 4103S - Potassium phosphate buffer

EMAIL THIS PAGE TO A FRIEND

4103S Sigma

Potassium phosphate buffer

0.1 M, pH 7.5 (25 °C), autoclaved

Purchase

Properties

sterility   autoclaved
clinical   for in vitro diagnostic use
IVD   for in vitro diagnostic use
concentration   0.1 M
pH   7.5 (25 °C)
storage temp.   2-8°C

Price and Availability

Customers Also Viewed

Potassium phosphate buffer solution

for HPLC, concentrate, ampule, pH 1.9

Potassium phosphate dibasic

ACS reagent, ≥98%

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Protocols & Articles

Related Products

replaced by

Product #

Description

Add to Cart

P5244 Phosphate buffer solution, 0.1 M

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?