• USA Home
  • 42491 - 1,2-Dioleoyl-sn-glycero-3-phosphoric acid sodium salt

EMAIL THIS PAGE TO A FRIEND
42491 Sigma

1,2-Dioleoyl-sn-glycero-3-phosphoric acid sodium salt

≥98.0% (TLC)

Synonym: 3-sn-Phosphatidic acid, 1,2-dioleoyl sodium salt, L-β,γ-Dioleoyl-α-phosphatidic acid sodium salt, DOPA

  • CAS Number 116004-31-0

  • Empirical Formula (Hill Notation) C39H71Na2O8P

  • Molecular Weight 744.93

  •  Beilstein Registry Number 3884561

  •  PubChem Substance ID 24866599

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerophosphates, Glycerophospholipid Metabolism, Glycerophospholipids,
assay   ≥98.0% (TLC)
solubility   chloroform: soluble20 mg/mL, clear, colorless
shipped in   wet ice
storage temp.   −20°C

Description

Biochem/physiol Actions

1,2-Dioleoyl-sn-glycero-3-phosphoric acid is a phosphatidic acid. Phospholipids, major membrane lipids, are derived from phosphatidic acid .

1,2-Dioleoyl-sn-glycero-3-phosphoric acid regulates phospholipid metabolic genes in yeast by binding to transcriptional factor, Opi1. This binding is regulated by a rapid decrease in pH in response to glucose starvation. 1,2-Dioleoyl-sn-glycero-3-phosphoric acid acts as intracellular pH biosensor, coupling membrane biogenesis to nutrient availability .

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?