Synonym: Biomolecule Stabilizer BioStab
Popular Documents: Datasheet (PDF)
| Related Categories | Antibody & Immunoassay Stabilizers, Biochemicals and Reagents, Biostabilization Solutions, Enzyme Stabilizers, General Biomolecule Stabilizers, |
| impurities | DNases, none detected |
| RNases, none detected | |
| phosphatases, none detected | |
| proteases, none detected | |
| suitability | in accordance for identity (HPLC) |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?