• USA Home
  • 43775 - 2,2′-Dithiodi(1-naphthylamine)

EMAIL THIS PAGE TO A FRIEND
43775 Sigma

2,2′-Dithiodi(1-naphthylamine)

for fluorescence, ≥97.0% (CHN)

Synonym: 1,1′-Diamino-2,2′-dinaphthyl disulfide, 2,2′-Dithio-bis(1-aminonaphthalene), DTAN

  • CAS Number 38262-57-6

  • Linear Formula (-SC10H6NH2)2

  • Molecular Weight 348.48

  •  Beilstein Registry Number 2886977

  •  MDL number MFCD00010499

  •  PubChem Substance ID 57650192

Purchase

Description

Other Notes

For the fluorimetric determination of aromatic aldehydes; For the determination of aldehydes used in the assay procedures for dopamine β-hydroxylase and catechol-O-methyltransferase

Application

2,2′-Dithiodi(1-naphthylamine) (DTAN) is used for the fluorimetric determination of aromatic aldehydes and for the determination of aldehydes used in the assay procedures for dopamine β-hydroxylase and catechol-O-methyltransferase.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?