• USA Home
  • 44608 - Durcupan water-soluble

EMAIL THIS PAGE TO A FRIEND
44608 Sigma

Durcupan water-soluble

single component D

Purchase

Description

Application

water-soluble embedding material for electron microscopy

Legal Information

Durcupan is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
TI0875000
Flash Point(F) 
339.8 °F
Flash Point(C) 
171 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?