• USA Home
  • 44617 - 6-Carboxy-X-rhodamine N-succinimidyl ester

EMAIL THIS PAGE TO A FRIEND
44617 Sigma

6-Carboxy-X-rhodamine N-succinimidyl ester

BioReagent, suitable for fluorescence

  • CAS Number 216699-36-4

  • Empirical Formula (Hill Notation) C37H33N3O7

  • Molecular Weight 631.67

  •  MDL number MFCD07437881

Purchase

Properties

Related Categories Biochemicals and Reagents, Detection, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Fluorescent Stains,
InChI Key   KWADUASJUMATIK-UHFFFAOYSA-N
product line   BioReagent
suitability   in accordance for fluorescence
  suitable for fluorescence
storage temp.   −20°C

Description

Other Notes

Real-time PCR for nucleic acid detection using labeled primers or nucleotides with this rhodamine X derivative

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Fluorescent Labeling of Peptides

Labeling peptides with fluorescent dyes or other labels provides powerful tools for the investigation of biological relevant interactions like receptor-ligand-binding,1-3 protein structures,4-6 and e...
ChemFiles Volume 5 Article 12
Keywords: Alzheimer Disease, Biochemistry, Electrophoresis, Enzyme activity, Immobilization, Ligands, Methods, Microscopy, Peptide synthesis, Separation, Solid phase peptide synthesis, Solvents, Spectroscopy, Tools

Related Products

replaced by

Product #

Description

Add to Cart

41969 5-Carboxy-X-rhodamine N-succinimidyl ester, BioReagent, suitable for fluorescence

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?