• USA Home
  • 45165 - EMPIGEN® BB detergent

EMAIL THIS PAGE TO A FRIEND
45165 Sigma

EMPIGEN® BB detergent

~35% active substance in H2O

Synonym: N,N-Dimethyl-N-dodecylglycine betaine, N-(Alkyl C10-C16)-N,N-dimethylglycine betaine

Purchase

Properties

Related Categories Antigen-Vaccine Preparation, Biochemicals and Reagents, Detergents, Detergents A to M, Detergents by Applications,
description   zwitterionic
InChI Key   DVEKCXOJTLDBFE-UHFFFAOYSA-N
mol wt   Mr ~279
concentration   ~35% active substance in H2O
CMC   1.6-2.1 mM(20-25°C)
density   1.04 g/mL at 20 °C
λ   neat
UV absorption   λ: 417 nm Amax: 0.055
  λ: 441 nm Amax: 0.037

Description

Application

Zwitterionic detergent (an amine betaine), useful for solubilization of membrane proteins. Used for preparing viral antigens by removing kinases.

Legal Information

EMPIGEN is a registered trademark of Albright & Wilson UK Ltd.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

30326 EMPIGEN® BB detergent, ~30% active substance

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?