• USA Home
  • 45712 - 2,4,5-T-1-octyl ester

EMAIL THIS PAGE TO A FRIEND
45712 Sigma

2,4,5-T-1-octyl ester

PESTANAL®, analytical standard

Synonym: Octyl (2,4,5-trichlorophenoxy)acetate

  • CAS Number 2630-15-1

  • Empirical Formula (Hill Notation) C16H21Cl3O3

  • Molecular Weight 367.70

  •  Beilstein Registry Number 2299251

  •  EC Number 220-109-9

  •  MDL number MFCD00128013

  •  PubChem Substance ID 329756921

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Pesticides & Metabolites Standards, Q-Z More...
grade   analytical standard
InChI Key   VXWISAHTAQYMSH-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?