• USA Home
  • 46064 - Esterase, immobilized on Eupergit® C from hog liver

EMAIL THIS PAGE TO A FRIEND
46064 Sigma

Esterase, immobilized on Eupergit® C from hog liver

~200 U/g moist material

Synonym: Esterase, immobilized on oxiran acrylicbeads from hog liver, PLE on Eupergit® C, Pig liver Esterase, immobilized on Eupergit® C,

Purchase

Description

General description

Immobilized on copolymer of methacrylamide, allyl-glycidylether and methylene-bis-acrylamide, moist pearls (dried material ~35%, pearl diameter ~150 μm)

Other Notes

Convenient reagent for the preparation of chiral building blocks

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol ethyl valerate (Cat. No. 30784) per minute at pH 8.0 and 25°C

Legal Information

Eupergit is a registered trademark of Röhm GmbH & Co. KG

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?