• USA Home
  • 46116 - 7-Ethoxy-4-methylcoumarin

EMAIL THIS PAGE TO A FRIEND
46116 Sigma

7-Ethoxy-4-methylcoumarin

suitable for fluorescence, ≥90% (TLC)

Synonym: Ethyl 4-methylumbelliferyl ether

  • CAS Number 87-05-8

  • Empirical Formula (Hill Notation) C12H12O3

  • Molecular Weight 204.22

  •  Beilstein Registry Number 169998

  •  EC Number 201-721-5

  •  MDL number MFCD00016971

  •  PubChem Substance ID 57650704

Purchase

Properties

Related Categories Biochemicals and Reagents, Detection, Fluorescent Enzyme Substrates, Fluorescent Enzyme Substrates for Spectroscopy, Fluorescent Probes, Labels, Particles and Stains,
InChI Key   NKRISXMDKXBVRJ-UHFFFAOYSA-N
assay   ≥90% (TLC)
mp   114-116 °C(lit.)
solubility   DMF: soluble
  acetonitrile: soluble
  alcohols: soluble
  chloroform: soluble
fluorescence   λex 318 nm; λem 377 nm in methanol(lit.)
  λex 360 nm; λem 449 nm in dealkylase(lit.)
suitability   suitable for fluorescence

Description

Application

7-Ethoxy-4-methylcoumarin is used as a substrate to identify and characterize coumarin 7-hydroxylase(s) and P450-isozymes. 7-Ethoxy-4-methylcoumarin may be used as a reference compound in analytical coumarin separation applications.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?