• USA Home
  • 47810 - D-Fructose 1,6-bisphosphate trisodium salt octahydrate

EMAIL THIS PAGE TO A FRIEND
47810 Sigma

D-Fructose 1,6-bisphosphate trisodium salt octahydrate

≥98.0% (NT)

Synonym: D-Fructose 1,6-diphosphate trisodium salt, FDP-Na3H

  • CAS Number 81028-91-3

  • Empirical Formula (Hill Notation) C6H11Na3O12P2 · 8H2O

  • Molecular Weight 550.18

  •  Beilstein Registry Number 3837985

  •  EC Number 253-778-0

  •  MDL number MFCD00149340

  •  PubChem Substance ID 57651116

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Monosaccharide More...
InChI Key   DBTJWGHAMYSWQR-MNDNZJPOSA-K
assay   ≥98.0% (NT)
impurities   ~25% water
storage temp.   2-8°C

Description

Application

D-Fructose-1,6-bisphosphate (FBP), a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1, 6-bisphosphatase(s). FBP is studied as a neuroprotective agent in brain injury.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Glycolysis Overview

Carbohydrates synthesized during photosynthesis act as the main storage molecules of solar energy. When ingested, complex carbohydrates are enzymatically hydrolyzed to monosaccharides, such as starch...
BioFiles 2007, 2.6, 4.
Keywords: Aldol reaction, Biofiles, Dehydration reaction, Glycolysis, Isomerizations, Metabolism, Metabolites, Metabolomics, Phosphorylations, Photosynthesis

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?