EMAIL THIS PAGE TO A FRIEND
48259 Sigma

D-(+)-Galactose

BioUltra, ≥99.5% (sum of enantiomers, HPLC)

Synonym: Galactose

  • CAS Number 59-23-4

  • Empirical Formula (Hill Notation) C6H12O6

  • Molecular Weight 180.16

  •  Beilstein Registry Number 1724619

  •  EC Number 200-416-4

  •  MDL number MFCD00151230

  •  PubChem Substance ID 57651152

Purchase

Properties

Related Categories Acacia, Agave sisalana, Allium sativum (Garlic), Aloe Vera, BioUltra,
InChI Key   GZCGUPFRVQAUEE-KCDKBNATSA-N
product line   BioUltra
assay   ≥99.5% (sum of enantiomers, HPLC)
optical activity   [α]20/D +80±1°, 24 hr, c = 5% in H2O
impurities   insoluble matter, passes filter test
ign. residue   ≤0.05% (as SO4)
loss   ≤0.05% loss on drying
pH   5.0-7.0 (25 °C, 1 M in H2O)
mp   168-170 °C(lit.)
solubility   H2O: soluble1 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
λ   1 M in H2O
UV absorption   λ: 260 nm Amax: 0.05
  λ: 280 nm Amax: 0.05

Description

Other Notes

Specifically inhibits agglutination of mouse spleen lymphocytes by soybean agglutinin

Application

D-(+)-Galactose (D-Gal) is a component with glucose of the disaccharide lactose. Galactose may be used in studies on galactose metabolism as the substrate for indentificaiton, differentiation and characterization of enzymes such as galactose oxidase(s) (GO), galactose permease(s), galactose kinase(s), and galactose phosphorylase(s). D-Gal is used in the study of Maillard glycation and its consequences in vivo.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
LW5490000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?