• USA Home
  • 49030 - γ-Globulins from bovine blood

EMAIL THIS PAGE TO A FRIEND
49030 Sigma

γ-Globulins from bovine blood

Cohn Fraction II, ≥95% (HPCE/GE)

Synonym: Bovine γ-Globulin

Purchase

Properties

Related Categories Biochemicals and Reagents, Bovine, Proteins and Derivatives More...
assay   ≥95% (HPCE/GE)
mol wt   Mr ~150000
loss   ≤5% loss on drying
solubility   0.05 M phosphate buffer pH 7.0: soluble10 mg/mL, hazy, colorless
storage temp.   2-8°C

Description

Application

Bovine γ-globulin has been used as an immunological challenge in studies of mechanisms of immunotolerance.

General description

Plasma protein fraction rich in antibodies

Other Notes

Review: Immunoglobulins

Sales restrictions may apply

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?