• USA Home
  • 49622 - L-Glutamic acid 1-(4-nitroanilide)

EMAIL THIS PAGE TO A FRIEND
49622 Sigma

L-Glutamic acid 1-(4-nitroanilide)

≥98.0% (T)

  • CAS Number 24032-35-7

  • Empirical Formula (Hill Notation) C11H13N3O5

  • Molecular Weight 267.24

  •  Beilstein Registry Number 3036815

  •  EC Number 245-994-9

  •  MDL number MFCD00038111

  •  PubChem Substance ID 24873020

Purchase

Properties

Related Categories Aminopeptidase A, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Substrates by Enzyme More...
InChI Key   TXKOUVRCDMSOJZ-LBPRGKRZSA-N
assay   ≥98.0% (T)
optical activity   [α]20/D +81.0±3.0°, c = 1% in 5 M HCl
storage temp.   2-8°C

Description

Other Notes

Chromogenic substrate for the assay (determination of liberated 4-nitroaniline at 410 nm) of aminopeptidase A (EC 3.4.11.7)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?