• USA Home
  • 49668 - Lipase A Candida antarctica, recombinant from Aspergillus oryzae

EMAIL THIS PAGE TO A FRIEND
49668 Sigma

Lipase A Candida antarctica, recombinant from Aspergillus oryzae

Purchase

Description

Biochem/physiol Actions

Lipase A from Candida Antarctica catalyzes hydrolysis of tertiary alcohols to form glycerol and fatty acids and shows selectivity for the N-acylation of β-amino esters.

Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.

Application

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TO9776500

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?