EMAIL THIS PAGE TO A FRIEND
50216 Sigma

Steareth-100

pellets, white

Synonym: Brij® 700, Polyethylene glycol 4400 octadecyl ether, Sympatens-AS

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents N to Z, Non-Ionic Detergents
description   non-ionic
form   pellets
impurities   ≤1% water
color   white
mp   56-60 °C

Description

Legal Information

Brij is a registered trademark of Croda International PLC

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
2
RTECS 
WI6300000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?