• USA Home
  • 50232 - Lewis-A trisaccharide 3′-sulfate sodium salt

EMAIL THIS PAGE TO A FRIEND
50232 Sigma

Lewis-A trisaccharide 3′-sulfate sodium salt

≥95.0% (TLC)

Synonym: Lea-sulfate sodium salt, SO3(O→l3)-β-D-Gal-(1→3)[α-L-Fuc-(1→4)]-D-GlcNAc, Sulfo-Lewis-A trisaccharide sodium salt

  • Empirical Formula (Hill Notation) C20H34NNaSO18

  • Molecular Weight 631.53

  •  PubChem Substance ID 329757581

Purchase

Properties

InChI Key   AYVQWBMHWUMCCB-LOBKVNCFSA-M
assay   ≥95.0% (TLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Sulfated tetrasaccharide which uniformly inhibits all three selectins with IC50 values in the low micromolar range.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?