• USA Home
  • 50985 - Guanidine thiocyanate solution

EMAIL THIS PAGE TO A FRIEND
50985 Sigma

Guanidine thiocyanate solution

BioUltra, for molecular biology, ~4 M in H2O

Synonym: Guanidine isothiocyanate, Guanidinium isothiocyanate, Guanidinium rhodanide, Guanidinium thiocyanate

  • CAS Number 593-84-0

  • Linear Formula NH2C(=NH)NH2 · HSCN

  • Molecular Weight 118.16

  •  Beilstein Registry Number 3563461

  •  MDL number MFCD00013027

  •  PubChem Substance ID 24873463

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Chaotropic Agents, DNA & RNA Purification, Denaturation,
grade   for molecular biology
InChI Key   ZJYYHGLJYGJLLN-UHFFFAOYSA-N
product line   BioUltra
concentration   ~4 M in H2O
impurities   DNases, none detected
  RNases, none detected
  insoluble matter, passes filter test
  phosphatases, none detected
  proteases, none detected
refractive index   n20/D 1.431
pH   4.5-7.0 (25 °C)
density   1.103 g/mL at 20 °C
anion traces   chloride (Cl-): ≤500 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Al: ≤1 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤1 mg/kg
  Bi: ≤1 mg/kg
  Ca: ≤5 mg/kg
  Cd: ≤1 mg/kg
  Co: ≤1 mg/kg
  Cr: ≤1 mg/kg
  Cu: ≤1 mg/kg
  Fe: ≤1 mg/kg
  K: ≤20 mg/kg
  Li: ≤1 mg/kg
  Mg: ≤1 mg/kg
  Mn: ≤1 mg/kg
  Mo: ≤1 mg/kg
  Na: ≤500 mg/kg
  Ni: ≤1 mg/kg
  Pb: ≤1 mg/kg
  Sr: ≤1 mg/kg
  Zn: ≤1 mg/kg
λ   neat
UV absorption   λ: 280 nm Amax: 0.7

Description

Caution

possibly crystallized product can be dissolved again by slightly heating to 40-50 C.

Other Notes

Extraction of total RNA

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Microcon Centrifugal Filters

All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN 3267 8 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?