• USA Home
  • 51251 - 6-(4-Acetamido-1,8-naphthalamido)hexanoic acid

EMAIL THIS PAGE TO A FRIEND
51251 Sigma

6-(4-Acetamido-1,8-naphthalamido)hexanoic acid

BioReagent, suitable for fluorescence, ≥95.0% (HPCE)

Synonym: 6-(4-Acetamino-1,8-naphthalamido)caproic acid, 6-(Acetamido)-1,3-dioxo-1H-benz[de]isoquinoline-2(3H)-hexanoic acid

  • CAS Number 172227-59-7

  • Empirical Formula (Hill Notation) C20H20N2O5

  • Molecular Weight 368.38

  •  Beilstein Registry Number 7443686

  •  MDL number MFCD04040049

  •  PubChem Substance ID 329757675

Purchase

Properties

Related Categories Biochemicals and Reagents, Derivatization agents, Luminescent Compounds/Detection More...
InChI Key   RCZAYEHQRFPFSH-UHFFFAOYSA-N
product line   BioReagent
assay   ≥95.0% (HPCE)
fluorescence   λex 364 nm; λem 476 nm in 0.1 M phosphate pH 7.0
suitability   suitable for fluorescence

Description

Other Notes

Fluorescent marker

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

6-(4-Acetamido-1,8-naphthalamido)hexanoic acid (6-(4-Acetamino-1,8-naphthalamido)caproic acid, NCA) is a fluorescent probe used for labeling antisense oligonucleotides.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?