• USA Home
  • 51553 - Heptaethylene glycol monodecyl ether

EMAIL THIS PAGE TO A FRIEND
51553 Sigma

Heptaethylene glycol monodecyl ether

≥97.0% (GC)

Synonym: C10E7, Decylheptaglycol, Polyoxyethylene (7) decyl ether

  • CAS Number 39840-09-0

  • Linear Formula CH3(CH2)9(OCH2CH2)7OH

  • Molecular Weight 466.65

  •  Beilstein Registry Number 2389035

  •  MDL number MFCD00043434

  •  PubChem Substance ID 57651329

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents A to M, Non-Ionic Detergents
description   non-ionic
InChI Key   DBJKLTWHVHVYJV-UHFFFAOYSA-N
assay   ≥97.0% (GC)
refractive index   n20/D 1.456
bp   300 °C
density   0.998 g/mL at 20 °C(lit.)
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cr: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤50 mg/kg
  Mg: ≤50 mg/kg
  Mn: ≤50 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?