• USA Home
  • 53718 - Hyaluronidase from bovine testes

EMAIL THIS PAGE TO A FRIEND
53718 Sigma

Hyaluronidase from bovine testes

90%, powder, yellow-brown, ~0.20 U/mg

Synonym: Hyaluronate 4-glycanohydrolase, Hyaluronoglucosaminidase

Purchase

Properties

Related Categories 3.2.x.x Glycosidases, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Carbohydrate Hydrolysis,
assay   90%
form   powder
color   yellow-brown
storage temp.   −20°C
Gene Information   cow ... HYAL1(515397)

Description

Biochem/physiol Actions

Hyaluronidase degrades hyaluronan and has been found to be inappropriately regulated during cancer progression.

These enzymes randomly cleave β-N-acetylhexosamine-[1→4] glycosidic bonds in hyaluronic acid, chondroitin, and chondroitin sulfates.

Other Notes

Sales restrictions may apply

Unit Definition

1 U corresponds to the amount of enzyme which releases 1 μmol N-acetyl-D-glucosamine end group per minute at pH 4.0 und 37°C (hyaluronic acid potassium salt; Cat. No. 53730, as substrate)

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?