• USA Home
  • 54832 - HiTrap® Blue Cartridge

EMAIL THIS PAGE TO A FRIEND
54832 Sigma

HiTrap® Blue Cartridge

part volume 1 mL

Synonym: HiTrap® Affinity Cartridge

Purchase

Properties

Related Categories Affinity Chromatography, Dye Resins, Molecular Biology, Protein Chromatography, Proteomics More...
packaging   pack of 5
mfr. no.   Amersham Biosciences HiTrap®, 17-0412-01
  Amersham Pharmacia Bio
part volume   1 mL
compatibility   mode of use Affinity
storage temp.   2-8°C

Description

General description

HiTrap adapters and instruction manual included.

Application

HiTrap® Blue Cartridges are used in affinity chromatography, protein chromatography and dye resins. The HiTrap® Blue albumin isolation system can be used for the analysis of albumin-organic toxicants adducts, identification of biomarkers in serum of early rheumatoid arthritis, and has been used to deplete plasma albumin for proteomic analysis of Bothrops jararaca snake plasma.

Legal Information

HiTrap is a registered trademark of GE Healthcare

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?