EMAIL THIS PAGE TO A FRIEND
56742 Sigma

Imbentin-AGS/35

25% in H2O

Synonym: Polyethylene glycol alkyl-(C11-C15) ether

  • Linear Formula HO(CH2CH2O)m(CH2)nCH3

  •  MDL number MFCD00212163

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Core Bioreagents, Detergents, Detergents A to M,
description   non-ionic
concentration   25% in H2O
refractive index   n20/D 1.366
density   1.005 g/mL at 20 °C(lit.)
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤100 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?