EMAIL THIS PAGE TO A FRIEND
60129 Sigma

Potassium chloride

BioUltra, ≥99.5% (AT)

Purchase

Properties

Related Categories Alphabetical, BioUltra Buffers, Biochemicals and Reagents, Biological Buffers, Molecular Biology,
InChI Key   WCUXLLCKKVVCTQ-UHFFFAOYSA-M
product line   BioUltra
assay   ≥99.5% (AT)
impurities   insoluble matter, passes filter test
  ≤0.001% total nitrogen (N)
loss   ≤0.2% loss on drying, 110 °C
pH   5.0-8.0 (25 °C, 1 M in H2O)
mp   770 °C(lit.)
solubility   H2O: soluble1 M at 20 °C, clear, colorless
density   1.98 g/mL at 25 °C(lit.)
anion traces   bromide (Br-): ≤500 mg/kg
  iodide (I-): ≤20 mg/kg
  phosphate (PO43-): ≤5 mg/kg
  sulfate (SO42-): ≤30 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤10 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤2 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤10 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Na: ≤200 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
λ   1 M in H2O
UV absorption   λ: 260 nm Amax: ≤0.01
  λ: 280 nm Amax: ≤0.01

Description

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.

Other Notes

Hypotonic KCl-propidium iodide protocol for univariate analysis of metaphase chromosomes (FCM); Potassium source for eukaryotic cell-free protein synthesizing systems; Dissociation of eukaryotic ribosomes into subunits by high concentrations of potassium (0.3-1.0 M); Preparation of mitochondria, provides an ionic osmotic support for those tissues that assume a gelatinous consistency upon homogenization.

Price and Availability


Amplified Detection

Interactive Buffer Table
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TS8050000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?