• USA Home
  • 61059 - Fluorescent red NIR 700 reactive

EMAIL THIS PAGE TO A FRIEND
61059 Sigma

Fluorescent red NIR 700 reactive

suitable for fluorescence

  • Empirical Formula (Hill Notation) C43H47N3O8S

  • Molecular Weight 765.91

  •  MDL number MFCD03792248

  • Popular Documents:  Datasheet (PDF)  

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Fluorescent Red and Orange Labels
solubility   DMF: soluble
fluorescence   λex 672 nm; λem ~735 nm in 0.1 M phosphate pH 7.0
suitability   suitable for fluorescence
storage temp.   −20°C

Description

Analysis Note

further Exc.: 700 nm (results in a lower emission)

Application

Fluorescent red 700 reactive is an amine conjugating fluorescent biolabel that can be used for covalent coupling to proteins and other biomolecules containing primary amino groups like amino modified DNA-oligomers and amino modified biotin. Fluorescent red 700 reactive is a new fluorescent label for the near infrared area. Extremely long wavelength fluorescence is especially well suited for applications where sample autofluorescence might be critical or penetration of tissue or other matrix is required.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?