• USA Home
  • 61309 - L-Lactic Dehydrogenase from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
61309 Sigma

L-Lactic Dehydrogenase from rabbit muscle

lyophilized, powder, off-white, ~140 U/mg

Synonym: L-LDH, L-Lactate Dehydrogenase from rabbit muscle, Lactate

Purchase

Properties

Related Categories 1.1.x.x Acting on hydroxyl groups, 1.x.x.x Oxidoreductases, Biochemicals and Reagents, Cell Biology, Chemical Synthesis,
form   powder
quality   lyophilized
mol wt   Mr ~140000
impurities   ≤0.01% glutamate pyruvate transaminase
  ≤0.01% malate Dehydrogenase
  ≤0.02% glutamate oxaloacetate transaminase
  ≤0.05% pyruvate kinase
color   off-white
storage temp.   2-8°C

Description

Biochem/physiol Actions

L-lactic dehydrogenase catalyzes the conversion of L-lactate into L-pyruvate while reducing NAD+ to NADH and H+.

Other Notes

In enzyme systems coupled to the NADH-dependent pyruvate-lactate reaction; Substrate specificity and use as a catalyst in the synthesis of homochiral 2-hydroxy acids; Inversion of chirality with an electroenzymatic reactor

Unit Definition

1 U corresponds to the amount of enzyme which reduces 1 μmol pyruvate per minute at pH 7.0 and 25 °C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?