EMAIL THIS PAGE TO A FRIEND
62323 Sigma

Lipase extension kit

Purchase

Description

Components

Composition:
62285 Lipase from Aspergillus oryzae, 100 mg
62288 Lipase B Candida antarctica, recombinant Aspergillus oryzae, 50mg
62304 Lipase from Mucor javanicus, 500 mg
62308 Lipase from Penicillium roqueforti, 500 mg
28602 Lipase from Pseudomonas fluorescens, 50 mg
90860 Lipase from Candida rugosa, 500mg
62306 Lipase from wheat germ, 500 mg
80612 Lipase from Rhizopus oryzae, 500 mg
62335 Lipoprotein Lipase from Pseudomonas sp., 10 mg
62336 Lipoprotein Lipase from Pseudomonas sp., 50 mg

Biochem/physiol Actions

Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?