• USA Home
  • 62639 - Lowicryl® K4M reagent kit

EMAIL THIS PAGE TO A FRIEND
62639 Sigma

Lowicryl® K4M reagent kit

for microscopy

Synonym: HMDSO

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Cell Biology, Embedding & Mounting Media Kits, Embedding and Mounting Media,
grade   for microscopy
impurities   methacrylates (not stabilized)
storage temp.   2-8°C

Description

Application

Polar, methacrylate/acrylate based, low- temperature embedding resin for biological material. Can be used with either the progressive lowering of temperature technique or freeze substitution.; Applicable temperature range, 220K to 340K

Legal Information

Lowicryl is a registered trademark of Polysciences

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
194 °F
Flash Point(C) 
90 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?