EMAIL THIS PAGE TO A FRIEND
64655 Sigma

Methocel® 60 HG

28-30% methoxyl basis

Synonym: Methyl cellulose

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Core Bioreagents, Polysaccharide, Research Essentials More...
assay   28-30% methoxyl basis
viscosity   35-55 mPa.s, 2 % in H2O(20 °C)

Description

Legal Information

Methocel is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?