• USA Home
  • 65221 - 1-(4-Methoxyphenyl)-6-phenylhexatriene

EMAIL THIS PAGE TO A FRIEND
65221 Sigma

1-(4-Methoxyphenyl)-6-phenylhexatriene

suitable for fluorescence, ≥98.0% (TLC)

Synonym: 4′-Methoxydiphenylhexatriene

  • CAS Number 3893-10-5

  • Empirical Formula (Hill Notation) C19H18O

  • Molecular Weight 262.35

  •  Beilstein Registry Number 1968057

  •  MDL number MFCD00674534

  •  PubChem Substance ID 57652022

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Detection, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains,
InChI Key   WSZUPMMXEJRMRA-SVRHENQESA-N
assay   ≥98.0% (TLC)
mp   196-198 °C(lit.)
solubility   alcohols: soluble
  chloroform: soluble
  toluene: soluble
fluorescence   λex 380 nm; λem 443 nm in chloroform
suitability   suitable for fluorescence

Description

Application

1-(4-Methoxyphenyl)-6-phenylhexatriene, a methyl derivative of 1,6-diphenyl-1,3,5-hexatriene (DPH), may be suitable for use as a Fluorescence Resonance Energy Transfer (FRET) pairing reagent and as a hydrophobic site labeling probe for membranes.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?