Synonym:
6-
Empirical Formula (Hill Notation) C30H33N3O9S
Molecular Weight 611.66
MDL number MFCD06798201
PubChem Substance ID 24885829
Popular Documents: Datasheet (PDF)
| Related Categories | Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Mega Labels |
| assay | ≥90% (coupling to amines) |
| fluorescence | λex 556 nm; λem 667 nm in 0.1 M phosphate pH 7.0 |
| suitability | suitable for fluorescence |
| storage temp. | 2-8°C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?