• USA Home
  • 68668 - Tracy 690 (activated)

EMAIL THIS PAGE TO A FRIEND
68668 Sigma

Tracy 690 (activated)

suitable for fluorescence

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Tracy Labels
solubility   DMF: soluble
fluorescence   λex 688 nm; λem 715 nm in 0.1 M phosphate pH 7.0
suitability   corresponds for coupling to amines
  suitable for fluorescence
storage temp.   2-8°C

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?