EMAIL THIS PAGE TO A FRIEND
69242 Sigma

Iris 7G-WS NHS ester

Synonym: 2-{2-{2-[4-(4-(2,5-Dioxopyrrolidinyloxycarbonyl)butyrylamino)phenylthio]-3-{2-[1-(hex-5-ynyl)-1,3-dihydro-3,3-dimethyl-5-sulfoindol-2-ylidene]ethylidene}cyclohex-1-enyl}vinyl}-1-(hex-5-ynyl)-3,3-dimethyl-3H-indolium-5-sulfonate monopotassium salt

  • Empirical Formula (Hill Notation) C57H61KN4O11S3

  • Molecular Weight 1113.41

  •  MDL number MFCD18452832

  •  PubChem Substance ID 329761494

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Other Fluorescent Labels
InChI Key   VKOGRNQHQLMODR-UHFFFAOYSA-M
fluorescence   λex 804 nm; λem 823 nm in 0.1 M phosphate pH 7.0
storage temp.   −20°C

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Principle

The NHS group conjugates to amino groups under mild condition and enables convenient labeling of proteins, antibodies, and other amine-containing biomolecules.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

New Products for Detection Applications

Isoelectric focusing (IEF) is a powerful analytical tool for the separation of proteins. In order to ensure the high performance of analysis, isoelectric point (pI) standards are needed. In addition ...
Keywords: Absorption, Applications, Bacterial conjugations, Capillary electrophoresis, Catalysis, Electrophoresis, Gel electrophoresis, Immobilization, Isoelectric focusing, Purification, Separation, Support

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?