• USA Home
  • 69782 - L-(+)-Lactic acid solution

EMAIL THIS PAGE TO A FRIEND
69782 Sigma

L-(+)-Lactic acid solution

purum, ~20% (L-lactic acid in water)

Synonym: (S)-2-Hydroxypropionic acid, Sarcolactic acid

  • CAS Number 79-33-4

  • Empirical Formula (Hill Notation) C3H6O3

  • Molecular Weight 90.08

  •  Beilstein Registry Number 1720251

  •  MDL number MFCD00064266

  •  PubChem Substance ID 57652209

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffer Convenience Packaging, Buffer Solutions, Carbohydrates,
InChI Key   JVTAAEKCZFNVCJ-REOHCLBHSA-N
grade   purum
assay   ~20% (L-lactic acid in water)
impurities   ≤1% D-lactic acid

Description

Other Notes

Aqueous solutions of l(+)-Lactate.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
1
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?