• USA Home
  • 69957 - Precision Protein Standards

EMAIL THIS PAGE TO A FRIEND
69957 Sigma

Precision Protein Standards

for electrophoresis

Purchase

Properties

Related Categories Molecular Biology, Molecular Weight Markers, Protein Electrophoresis, Proteomics
grade   for electrophoresis
suitability   in accordance for SDS-PAGE
storage temp.   −20°C

Description

Components

10 recombinant proteins, prestained (10, 15, 20, 25, 37, 50, 75, 100, 150, 250 kDa). 0.65 mg protein in 500 μl 30 % (v/v) glycerol, 2% SDS, 62.5 mM Tris, pH 6.8, 10 mM DTT, 5 mM EDTA, 0.02 % NaN3. Sufficient for 50 applications.

General description

Ten specially engineered proteins purified to yield bands of unrivaled clarity and sharpness, no lot-to-lot variation, preblended with sample loading buffer for optimal resolution and convenience.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Flash Point(F) 
320 °F
Flash Point(C) 
160 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?