• USA Home
  • 70495 - Naphthol AS-MX phosphate disodium salt nonahydrate

EMAIL THIS PAGE TO A FRIEND
70495 Sigma

Naphthol AS-MX phosphate disodium salt nonahydrate

for histology, ≥99.0% (TLC)

  • CAS Number 36889-52-8

  • Empirical Formula (Hill Notation) C19H16NNa2O5P · 9H2O

  • Molecular Weight 577.42

  •  MDL number MFCD00150898

  •  PubChem Substance ID 57652252

Purchase

Properties

Related Categories Acid Phosphatase, Alkaline Phosphatase, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates,
grade   for histology
InChI Key   GEMAPHRVLCZBIQ-UHFFFAOYSA-L
assay   ≥99.0% (TLC)
solubility   H2O: soluble0.1 g/mL, clear, light yellow
suitability   suitable as substrate for alkaline phosphatase test
storage temp.   −20°C

Description

Other Notes

Histochemical detection of alkaline phosphatase.; Enzyme-incorporated erythrocyte ghosts

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?