Synonym:
KLH,
Keyhole-
MDL number MFCD02687589
| Related Categories | Cell Culture, Other adjuvants, Vaccine Adjuvants, Vaccine Production |
| quality | solution in phosphate buffered saline, pH 7.2 |
| contains | 15 mM sodium azide as preservative |
| concentration | ~6 mg/mL protein |
| impurities | 0.5 mM MgCl2 |
| 1 mM CaCl2 | |
| storage temp. | 2-8°C |
Carrier protein, use as antigen1,2
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?