EMAIL THIS PAGE TO A FRIEND
71184 Sigma

Sodium acetate

BioUltra, for luminescence, anhydrous, ≥99.0% (NT)

Synonym: Acetic acid sodium salt

Purchase

Properties

Related Categories Alphabetical, BioUltra Buffers, Biochemicals and Reagents, Biological Buffers, Luminescent Compounds/Detection,
grade   for luminescence
InChI Key   VMHLLURERBWHNL-UHFFFAOYSA-M
product line   BioUltra
assay   ≥99.0% (NT)
form   solid
autoignition temp.   1112 °F
impurities   insoluble matter, passes filter test
loss   ≤0.5% loss on drying, 150 °C
pH   8.5-10.0 (25 °C, 3 M in H2O)
pKa    4.76 (acetic acid)
mp   >300 °C (dec.)(lit.)
solubility   H2O: soluble3 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): ≤20 mg/kg
  phosphate (PO43-): ≤5 mg/kg
  sulfate (SO42-): ≤20 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤10 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤200 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤10 mg/kg
  Zn: ≤5 mg/kg
λ   3 M in H2O
UV absorption   λ: 260 nm Amax: 0.03
  λ: 280 nm Amax: 0.02
suitability   in accordance for luminescence

Price and Availability


Laboratory Gloves

Amplified Detection

Interactive Buffer Table
Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
AJ4300010
Flash Point(F) 
>482 °F
Flash Point(C) 
>250 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?