• USA Home
  • 71603 - Bis(2,2′-bipyridine)-(5-isothiocyanato-phenanthroline)ruthenium bis(hexafluorophosphate)

EMAIL THIS PAGE TO A FRIEND
71603 Sigma

Bis(2,2′-bipyridine)-(5-isothiocyanato-phenanthroline)ruthenium bis(hexafluorophosphate)

BioReagent, suitable for fluorescence

Synonym: Ru(bpy)2(phen-5-isothiocyanate)(PF6)2

  • CAS Number 288399-07-5

  • Empirical Formula (Hill Notation) C33H23F12N7P2RuS

  • Molecular Weight 940.65

  •  MDL number MFCD02683413

  •  PubChem Substance ID 57652323

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Luminescent Compounds/Detection, Metal Complexes,
InChI Key   VUUXKQLUOQLWPZ-UHFFFAOYSA-N
product line   BioReagent
fluorescence   λex 445 nm; λem ~615 nm in 0.1 M phosphate pH 7.0
suitability   suitable for fluorescence
shipped in   dry ice
storage temp.   −20°C

Description

Application

Reactive probe containing fluorescent metal-ligand complex for determination of nucleotide base sequences.

Features and Benefits

Sigma-Aldrich offers a series of reactive dyes based on ruthenium complexes for use in fluorescence polarization assays and fluorescence lifetime imaging applications. They have been designed specifically to have long fluorescence lifetimes. These dyes were first synthesized in the labs of Dr. Joseph Lakowicz at the Center for Fluorescence Microscopy. These laser-excitable metal ligand complexes have fluorescence lifetimes near 500 nanoseconds. These dyes can be used to label amines on biomolecules under mild conditions. Advantages of these dyes as protein labels include high photostability, good water solubility, a lack of dye-dye interactions and large Stokes’ shifts. In addition, the fluorescence signal of long-lived fluorophores can be gated to eliminate the emission from short-lifetime fluorophores and autofluorescence from cells and biomolecules to further improve sensitivity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?