• USA Home
  • 72674 - 1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, pyrene-labeled

EMAIL THIS PAGE TO A FRIEND
72674 Sigma

1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, pyrene-labeled

BioReagent, suitable for fluorescence, ≥98.0% (TLC)

Synonym: L-β,γ-Dioleoyl-α-cephaline, pyrene-labeled, N-(1-Pyrenesulfonyl)-1,2-dioleoyl-sn-glycero-3-phosphoethanolamine, Phosphatidylethanolamine 18:1, pyrene-labeled

  • Empirical Formula (Hill Notation) C57H86NO10PS

  • Molecular Weight 1008.33

  •  MDL number MFCD02259237

  •  PubChem Substance ID 57652430

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains, Fluorescent labeled lipids for membrane probes,
InChI Key   VQMRCQVKHOAKOB-YQAYVEMKSA-N
product line   BioReagent
assay   ≥98.0% (TLC)
fluorescence   λex 323 nm; λem 378 nm in methanol
suitability   suitable for fluorescence
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Sensor Dyes

In 1998 R.F. Furchgott, L.J. Ignarro, and F. Murad received the Nobel Prize in Physiology and Medicine for their discoveries concerning nitric oxide as a signaling molecule in the cardiovascular syst...
Monika Baeumle
BioFiles 2011, 6.3, 23.
Keywords: Applications, Biochemistry, Biological processes, Cancer, Cardiovascular, Central Nervous System, Diseases, Extinction coefficient, Indicators, Inflammation, Methods, Microscopy, Neurotransmission, Sol-gel, Type

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?