• USA Home
  • 73038 - InP/ZnS nanoparticles

EMAIL THIS PAGE TO A FRIEND
73038 Sigma

InP/ZnS nanoparticles

core-shell type, yellow emitting, 1 mg/mL

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Nanoparticles, Fluorescent Probes, Labels, Particles and Stains More...
concentration   1 mg/mL
fluorescence   λem 565 nm±20 nm

Description

Analysis Note

full width at half maximum ≤100 nm

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1294 - class 3 - PG 2 - Toluene, solution
WGK Germany 
2
Flash Point(F) 
39.2 °F
Flash Point(C) 
4 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?