EMAIL THIS PAGE TO A FRIEND
73248 Sigma

Melengestrol acetate

≥97.0% (HPLC)

Synonym: 17α-Acetoxy-6-methyl-16-methylene-4,6-pregnadiene-3,20-dione, MGA

  • CAS Number 2919-66-6

  • Empirical Formula (Hill Notation) C25H32O4

  • Molecular Weight 396.52

  •  Beilstein Registry Number 2066919

  •  EC Number 220-859-7

  •  MDL number MFCD00072142

  •  PubChem Substance ID 57652468

Purchase

Properties

InChI Key   UDKABVSQKJNZBH-DWNQPYOZSA-N
assay   ≥97.0% (HPLC)
mp   202-204 °C(lit.)

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Biochem/physiol Actions

Causes shrinkage of androgen-dependent prostate tumors.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TU4141000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?